missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ TYK2 Recombinant Human Protein DFS Item

Tyrosine Kinase 2 (TYK2) is a member of the JAK kinase family

Manufacturer:  Invitrogen™ LSPV4790

 View more versions of this product

Catalog No. PV4790



Description

Description

Tyrosine Kinase 2 (TYK2) a member of the JAK kinase family, plays both structural and catalytic roles in type I interferon (IFN) signaling.
Specifications

Specifications

NP_003322.2
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
10 µg
LanthaScreen® Antibody, LanthaScreen® Substrate, Z'-LYTE™
TYK2
Baculovirus
Recombinant, Cytoplasmic, Catalytic
Signal Transduction
Kinase Assay
68.8 kDa
JAK Kinase Family
GST Tag
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Tyrosine Kinases
Dry Ice
N-Terminal
tyrosine kinase 2
Kinases & Inhibitors
Cytoplasmic Tyrosine Kinases
Human
7297
TYK2
aa 833-1187
Documents