Saint-Gobain™ Tygon™ Biocompatible Tubing, Formula S-50-HL
Tubing ends closed by heat-sealing to prevent contamination in shipment
$31.55 - $31.55
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
14-168-111 Accessories Saint Gobain Perform Plas
ADF00002
Pack for $31.55
Specifications
Description
  • Crystal clear for easy visualization of fluid flow
  • Nonwetting surface permits complete drainage
  • Greater flex-life than rubber
  • Can be autoclaved or sterilized with steam, radiation, chemicals, or gas
  • Durometer hardness: Shore A, 66
Order Info
In pack of 50 ft. (15.2m)
Compliance
Meets U.S. Pharmacopeia Class VI criteria for medical plastics. Complies with CID A-A-55697 for medical tubing. Follows ISO 10993 standards.