Thermo Scientific™ TX-750 Swinging Bucket/BIOShield 1000A Adapters
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
Supplier:  Thermo Scientific™ 75003749
 View more versions of this product
Catalog No. 75-003-749
Description
Description
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
  • Maximum tube size: 63 × 130mm
Specifications
Specifications
50mL Round-bottom Tube