Please login to your online account to display your discounted pricing

S-Curve™ Two Compartment Gloves Dispenser

$402.18 - $402.18

Specifications

Color Clear
Product Type Glove Dispenser
For Use With (Application) Sloped top to maintain HEPA flow
No. of compartments 2
View More Specs
Products
Catalog Number Mfr. No. Material Price Quantity    

19-170-652

 
s-curve™
SD200*
Acrylic Each for $402.18
Description & Specifications

Specifications

Color Clear
Product Type Glove Dispenser
For Use With (Application) Sloped top to maintain HEPA flow
No. of compartments 2