Please login to your online account to display your discounted pricing

Twistit: Four Stoppers in One

Multi-purpose and economical

$31.55 - $31.55

Specifications

Color White
Material Rubber
Finish Clinical
For Use With (Application) Multi-purpose applications
View More Specs
Products
Catalog Number Mfr. No. Size Stopper Number Product Type Price Quantity    

S35081

 
fisher science education
ZM669ASST
3 to 9 Assorted Twistit™ Stopper Pack of 33 Each for $31.55
Description & Specifications

Specifications

Color White
Material Rubber
Finish Clinical
For Use With (Application) Multi-purpose applications

Four stoppers in one! Seals twist off to adapt the stopper to your application. Use as a solid, one-, two-, or three-hole stopper. Reduces inventory, confusion, and cost. Made of clean, white, resilient, natural rubber with a clinical finish.