missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Inc Tween™80, ∽75% oleic acid, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210317091

 View more versions of this product

Catalog No. ICN10317091



Description

Description

Specifications

Specifications

Tween 80
9005-70-3
1.08g/mL
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
1 L
ZGEQUSWSYZZNAI-CLFAGFIQNA-N
NaN
Yellow
Liquid
∽75%
MFCD01779641
Polysorbate 80, Polyoxyethylene sorbitan monooleate
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO

Chemical Identifiers

9005-70-3
NaN
ZGEQUSWSYZZNAI-CLFAGFIQNA-N
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
MFCD01779641
Polysorbate 80, Polyoxyethylene sorbitan monooleate
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions