missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polysorbate 40, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 334140010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polysorbate 40 | |
9005-66-7 | |
1.0500g/mL | |
MFCD00165345 | |
Polyoxyethylene (20) Sorbitan Monopalmitate, Polysorbate 40 | |
>200°C | |
CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
89 to 105mg KOH/g | |
92329579 | |
Glass bottle | |
1.05 |
Yellow | |
Viscous Liquid or Paste At 25°C | |
(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
1 L | |
Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, xylene, insoluble in propylene glycol and mineral oil | |
DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
522.68 | |
Authentic | |
41 to 52mg KOH/g | |
ca.200 mPa.s (50°C) |
Chemical Identifiers
9005-66-7 | |
522.68 | |
DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
92329579 |
(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
MFCD00165345 | |
Polyoxyethylene (20) Sorbitan Monopalmitate, Polysorbate 40 | |
CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
RUO – Research Use Only