missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ TWEEN™ 20, Protein Grade™ Detergent, 10% Solution, Sterile-Filtered, Calbiochem™,
Manufacturer: MilliporeSigma™ 65520650ML
Specifications
9005-64-5 | |
Polyoxyethylene (20) sorbitan Monolaurate | |
HMFKFHLTUCJZJO-UHFFFAOYSA-N | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
443314 | |
50mL |
C26H50O10 | |
Glass Bottle | |
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO | |
522.676 | |
Protein |
Chemical Identifiers
9005-64-5 | |
522.676 | |
Polyoxyethylene (20) sorbitan Monolaurate | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate |
C26H50O10 | |
HMFKFHLTUCJZJO-UHFFFAOYSA-N | |
443314 | |
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO |