missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polysorbate 20, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 233362500

 View more versions of this product

Catalog No. AC233362500



Description

Description

Specifications

Specifications

Polysorbate 20
9005-64-5
10ppm max.
C26H50O10
Authentic
Glass Bottle
HMFKFHLTUCJZJO-UHFFFAOYSA-N
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
443314
Liquid
400 mPa.s (25°C)
>100°C
>150°C
1.1
2mg KOH/g max.
1.1000g/mL
96 to 108mg KOH/g
MFCD00165986
Polyoxyethylene(20)sorbitan monolaurate
Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
522.676
1.4685 to 1.4715
40 to 50mg KOH/g
3% Max.
Colorless to Yellow
250mL

Chemical Identifiers

9005-64-5
522.676
HMFKFHLTUCJZJO-UHFFFAOYSA-N
443314
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
C26H50O10
MFCD00165986
Polyoxyethylene(20)sorbitan monolaurate
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Safety and Handling

Safety and Handling

RTECSNumber : TR7400000

TSCA : TSCA

SDS Documents

RMF - Research Use or Further Mfg