Please login to your online account to display your discounted pricing

TW 37, Tocris™

Bcl-2 inhibitor; induces apoptosis

$297.57 - $1,231.36

Chemical Identifiers

Chemical Name or Material TW 37
Assay Percent Range >97%
Molecular Formula C33H35NO6S
Formula Weight 573.7
Synonym N-[4-[[2-(1,1-Dimethylethyl)phenyl]sulfonyl]phenyl]-2,3,4-trihydroxy-5-[[2-(1-methylethyl)phenyl]methyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

403810

 
r & d systems
RDS403810
10mg Each for $297.57

403850

 
r & d systems
RDS403850
50mg Each for $1,231.36
Description & Specifications

Specifications

Formula Weight 573.7
Quantity 10mg
Assay Percent Range >97%
Recommended Storage Store at -20°C
Molecular Formula C33H35NO6S
CAS 877877-35-5
Chemical Name or Material TW 37
Synonym N-[4-[[2-(1,1-Dimethylethyl)phenyl]sulfonyl]phenyl]-2,3,4-trihydroxy-5-[[2-(1-methylethyl)phenyl]methyl]benzamide
Safety and Handling

Store at -20°C