Please login to your online account to display your discounted pricing

Thermo Scientific™ TurboFect Transfection Reagent


Get efficient in vitro delivery of plasmid DNA to cultured cells with low cytotoxicity.

Manufacturer: thermo scientific™  R0541

 View more versions of this product

Catalog No. FERR0541

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) DNA transfection.siRNA-DNA co-transfection.
Quantity 120µL
Quantity 120µL
Sufficient For 20 transfections using 50µg DNA

TurboFect Transfection Reagent is recommended for efficient transfection of plasmid DNA. Distinct from lipid-based formulas, TurboFect is a sterile solution of a proprietary cationic polymer in water. The polymer forms positively-charged complexes with DNA that are both compact and stable. These complexes protect DNA from degradation and facilitate efficient plasmid delivery into eukaryotic cells.

Highlights

  • High transfection efficiency of plasmid DNA into a wide variety of cell types
    • Adherent and suspension cells
    • Differentiated and undifferentiated cells
  • Excellent transfection efficiency in the presence or absence of serum 
  • Ready-to-use reagent - no need to reconstitute, dilute, or manipulate
  • Simple protocol

Review our experimental considerations to improve your plasmid transfection results.

Review our experimental considerations to improve your plasmid transfection results.