missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Turbidity Stock Solution II, Hexamethylene Tetramine, 10%, LabChem™

Manufacturer:  Lab Chem LC263201

 View more versions of this product

Catalog No. LC263201



Specifications

Specifications

Turbidity Stock Solution II
100-97-0,7732-18-5
C6H12N4
Passes Test
10%
Soluble in water
Colorless
Hexamethylene Tetramine
90,10
140.19
Liquid
Poly Bottle
Gaseous Ammonia; Nitrogen oxides
500mL
Documents
Promotions

Promotions