Please login to your online account to display your discounted pricing

Turbidity Standard, 4000 NTU, Certified, LabChem™

$115.71 - $115.71

Chemical Identifiers

Chemical Name or Material Turbidity Standard
CAS 7732-18-5
CAS Max % 94.5
CAS 100-97-0
CAS Max % 5
CAS 10034-93-2
CAS Max % 0.5
Name Note 4000 NTU
Purity Grade Certified Grade
Linear Formula H4N2·H2SO4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC262901

 
lab chem
LC262901
Poly Bottle 500mL Each for $115.71
Description & Specifications

Specifications

Formula Weight 130.12
Purity Grade Certified Grade
Synonym Formazin
Physical Form Liquid
CAS Max % 94.5
CAS 7732-18-5
Name Note 4000 NTU
CAS Max % 0.5
Color Turbid, colorless
CAS Max % 5
Quantity 500mL
Packaging Poly Bottle
Linear Formula H4N2·H2SO4
Health Hazard 2 GHS H Statement
May cause an allergic skin reaction.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Solubility Soluble in water
Recommended Storage Room Temperature; Mix well before use
Health Hazard 1 Danger
CAS 10034-93-2
CAS 100-97-0
Chemical Name or Material Turbidity Standard
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear eye protection, protective gloves.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear eye protection, protective gloves.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause an allergic skin reaction.
May cause cancer.
Harmful to aquatic life with long lasting effects.

Room Temperature; Mix well before use