missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Turbidity Standard, 1000 NTU (Nephelometric Turbidity Units), Ricca Chemical

$130.00 - $130.00

Chemical Identifiers

CAS 7732-18-5,100-97-0,10034-93-2
CAS Min % 97.0,0.94,0.09
CAS Max % 100.0,0.98,0.1
Physical Form Suspension
Chemical Name or Material Turbidity Standard, 1000NTU (Nephelometric Turbidity Units)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
8825-16 SDS Ricca Chemical Company
RI882516
Glass amber 500mL Each for $130.00
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,100-97-0,10034-93-2
100.0,0.98,0.1
Turbidity Standard, 1000NTU (Nephelometric Turbidity Units)
97.0,0.94,0.09
Suspension

Specifications

Suspension
500mL
0.09,97.0,0.94
Glass amber
Hazy to milky white
7732-18-5,10034-93-2,100-97-0
0.98,0.1,100.0
Turbidity Standard, 1000NTU (Nephelometric Turbidity Units)