missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Turbidity Standard, 100 NTU (Nephelometric Turbidity Units) - Polymer Bead Suspension, Ricca Chemical

$388.09 - $1,255.97

Chemical Identifiers

CAS 7732-18-5,9003-53-6,26628-22-8
CAS Min % 98.0,0.18,0.01
CAS Max % 100.0,0.19,0.01
Physical Form Liquid
Chemical Name or Material Turbidity Standard, 100NTU (Nephelometric Turbidity Units) - Polymer Bead Suspension
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
88061 Ricca Chemical Company
RI88061
Glass amber 4L Each for $1,255.97
88-061-6 SDS Ricca Chemical Company
RI880616
PETG clear 500mL Each for $388.09
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,9003-53-6,26628-22-8
100.0,0.19,0.01
Turbidity Standard, 100NTU (Nephelometric Turbidity Units) - Polymer Bead Suspension
98.0,0.18,0.01
Liquid

Specifications

Liquid
4L
0.18,0.01,98.0
Glass amber
Clear to slightly hazy, colorless
9003-53-6,26628-22-8,7732-18-5
100.0,0.19,0.01
Turbidity Standard, 100NTU (Nephelometric Turbidity Units) - Polymer Bead Suspension