missing translation for 'onlineSavingsMsg'
Learn More

Tunicamycin, 10 mg/ml in DMSO, sterile-filtered

Catalog No. AAJ67401XF
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67401XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67401XF Supplier Thermo Scientific Chemicals Supplier No. J67401XF

CAS: 11089-65-9

Tunicamycin, inhibits the expression of functional thrombin receptors on human T-lymphoblastoid cells. Also inhibits thrombin-induced Ca2+ mobilization in cells. Also available as a 25 mM solution in DMSO. Antibacterial and antifungal. Blocks the formation of protein N-glycosidic linkages

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11089-65-9
MDL Number MFCD00065709

Specifications

Type Tunicamycin
Physical Form Liquid
Packaging Plastic Bottle
Quantity 1 mL
Solubility Information Soluble in DMF,DMSO,or Pyridine.
Chemical Name or Material Tunicamycin
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
RTECSNumber YO7980200
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only