Please login to your online account to display your discounted pricing

Tungsten wire, Au 99.99%, Alfa Aesar™

Form: approx. 0.005g/m (W only), 3-5 wt% Au plating

Chemical Identifiers

Chemical Name or Material Tungsten
Physical Form Wire, Gold Plated, 0.025mm (0.001 in.) dia.
Assay Percent Range 99.99% (Metals basis)
MDL Number MFCD00011461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41038G1

 
alfa aesar
41038G1
1m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41038G5

 
alfa aesar
41038G5
5m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41038G6

 
alfa aesar
41038G6
10m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Wire, Gold Plated, 0.025mm (0.001 in.) dia.
Quantity 1m
Color Gold colored
Chemical Name or Material Tungsten
Assay Percent Range 99.99% (Metals basis)
MDL Number MFCD00011461
Odor Odorless
Appearance Gold colored wire
Density 19.3g/cm3