missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten wire, 1.0mm dia., 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 10411G5

 View more versions of this product

Catalog No. AA10411G5



Specifications

Specifications

Tungsten
7440-33-7
19.3g/cm3
W
4+ ion
[W]
183.84
CHEBI:27998
5m
Silver Gray wire
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire, 1.0mm (0.04 in.) dia.

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

Documents