missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten wire, 99.95% (metal basis), Alfa Aesar™

Form: approx. 0.003g/m

Manufacturer:  Alfa Aesar 14512H9

 View more versions of this product

Catalog No. AA14512H9



Specifications

Specifications

Tungsten
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire, 0.015mm (0.00059 in.) dia.
500m
Silver Gray wirw
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Silver to Gray

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

SDS Documents