missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten wire, 1.4mm (0.055 in.) dia., 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 10412G1

 View more versions of this product

Catalog No. AA10412G1



Specifications

Specifications

Tungsten wire
1m
7440-33-7
W
4+ ion
[W]
183.84
CHEBI:27998
99.95%
≈29.7g/m
1.4mm (0.055 in.) dia.
(metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents