missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten Wire, 0.375mm (0.015 in.) dia., annealed, 99.95% (Metals basis), Alfa Aesar™

Form: Approx. 2.13g/m

Manufacturer:  Alfa Aesar 43995G5

 View more versions of this product

Catalog No. AA43995G5



Specifications

Specifications

Tungsten
Annealed
Silver Gray wire
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Silver to Gray
5m
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire, 0.375mm (0.015 in.) dia.

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]
Documents