missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten Wire, 0.1mm (0.004 in.) dia., 99.95% (Metals basis), Alfa Aesar™

Form: Approx. 0.15g/m

Manufacturer:  Thermo Fisher Scientific 10404I6

 View more versions of this product

Catalog No. AA10404I6



Specifications

Specifications

Tungsten
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire, 0.1mm (0.004 in.) dia.
4 x 250m
Silver Gray wire
W
4+ ion
[W]
183.84
CHEBI:27998
Silver to Gray

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Documents