missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten Wire, 0.025mm (0.001 in.) dia., 99.95% (Metals basis), Alfa Aesar™

Form: Approx. 0.009g/m

Manufacturer:  Alfa Aesar 10405H4

 View more versions of this product

Catalog No. AA10405H4



Specifications

Specifications

Tungsten
7440-33-7
19.3g/cm3
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Silver to Gray
100m
Silver Gray
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire, 0.025mm (0.001 in.) dia.

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]
Documents