missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten(VI) oxide, 99.8% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1182836

 View more versions of this product

Catalog No. AA1182836



Specifications

Specifications

Tungsten(VI) oxide
99.8% (metals basis)
MFCD00011466
ZNOKGRXACCSDPY-UHFFFAOYSA-N
trioxotungsten
14811
10 To 20 Micron Powder
500g
1314-35-8
O3W
tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
O=[W](=O)=O
231.837
231.85
7.16g/mL

Chemical Identifiers

1314-35-8
231.837
ZNOKGRXACCSDPY-UHFFFAOYSA-N
14811
O=[W](=O)=O
O3W
MFCD00011466
tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
trioxotungsten
Safety and Handling

Safety and Handling

EINECSNumber : 215-231-4

Documents