Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten(VI) isopropoxide, 5% w/v in isopropanol

Precursor for Tungsten oxide using sol-gel process

Chemical Identifiers

Chemical Name or Material Tungsten(VI) isopropoxide
CAS 52321-90-1
Linear Formula W[OCH(CH3)2]6
Formula Weight 538.38g/mol
MDL Number MFCD00210652
UN Number UN1219
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40247AC

 
alfa aesar™
40247-AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40247AE

 
alfa aesar™
40247-AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25mL
Concentration 5% w/v in isopropanol
Quantity 25mL
Sensitivity Moisture Sensitive
Chemical Name or Material Tungsten(VI) isopropoxide
CAS 52321-90-1
Physical Form Liquid
Linear Formula W[OCH(CH3)2]6
Formula Weight 538.38g/mol
MDL Number MFCD00210652
UN Number UN1219
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.