missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten(VI) ethoxide, Thermo Scientific Chemicals

Intermediate for sol-gel coatings

Supplier:  Thermo Scientific Chemicals 04179506

 View more versions of this product

Catalog No. AA4179506


Description

Description

Intermediate for sol-gel coatingsTungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. Also, it acts as an intermediate for sol-gel coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tungsten(VI) ethoxide
150°C
C12H30O6W
MFCD00156526
UN3181
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide
JHZFYDXXWXXVRM-UHFFFAOYSA-N
ethanolate;tungsten
13828235
Chunks and/or Powder
62571-53-3
White to Brown
W(OC2H5)6
5 g
Moisture sensitive
Soluble in ethyl acetateSoluble in ethyl acetate,hydrocarbons and esters.
CCO[W](OCC)(OCC)(OCC)(OCC)OCC
454.21
454.21

Chemical Identifiers

62571-53-3
454.21
JHZFYDXXWXXVRM-UHFFFAOYSA-N
13828235
CCO[W](OCC)(OCC)(OCC)(OCC)OCC
C12H30O6W
MFCD00156526
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide
ethanolate;tungsten
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only