missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten(VI) Chloride, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1233730

 View more versions of this product

Catalog No. AA1233730



Specifications

Specifications

Tungsten(VI) Chloride
99%
MFCD00011463
KPGXUAIFQMJJFB-UHFFFAOYSA-H
hexachlorotungsten
83301
396.57
Moisture sensitive
13283-01-7
Cl6W
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
396.54
CHEBI:37771
Crystalline
250g

Chemical Identifiers

13283-01-7
396.54
KPGXUAIFQMJJFB-UHFFFAOYSA-H
83301
hexachlorotungsten
Cl6W
MFCD00011463
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
CHEBI:37771
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

Causes burns.

DOTInformation : DOT Class: 8

EINECSNumber : 236-293-9

Documents