missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten(VI) chloride, 99%, 20 mesh, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 200302500

Catalog No. AC200302500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tungsten(VI) chloride
13283-01-7
99%
WCl6
04,569; 05,742; 06,658; 14,346; 09,498; 10,444; 12,566; 13,343
Solubility in water: decomposes. Other solubilities: soluble in cs2,chloroform,methyl alcohol,ether,,acetone,pyridine,glycerol,ligroin,ethanol,,carbon tetrachloride,phosphorus oxytrichloride,,benzene
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
396.57
CHEBI:37771
99%
20 Mesh
250g
Cl6W
MFCD00011463
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
KPGXUAIFQMJJFB-UHFFFAOYSA-H
hexachlorotungsten
83301
396.57
Glass bottle
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 236-293-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only