Learn More
Tungsten(VI) chloride, 99%, 20 mesh, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 200300500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tungsten(VI) chloride | |
13283-01-7 | |
99% | |
WCl6 | |
04,569; 05,742; 06,658; 14,346; 09,498; 10,444; 12,566; 13,343 | |
Solubility in water: decomposes. Other solubilities: soluble in cs2,chloroform,methyl alcohol,ether,,acetone,pyridine,glycerol,ligroin,ethanol,,carbon tetrachloride,phosphorus oxytrichloride,,benzene | |
Cl[W](Cl)(Cl)(Cl)(Cl)Cl | |
396.57 | |
CHEBI:37771 | |
99% |
20 Mesh | |
50g | |
Cl6W | |
MFCD00011463 | |
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten | |
KPGXUAIFQMJJFB-UHFFFAOYSA-H | |
hexachlorotungsten | |
83301 | |
396.57 | |
Glass bottle |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 236-293-9
RUO â Research Use Only