missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten(V) ethoxide 1,2-dimethoxyethane adduct, 99%

Manufacturer:  Alfa Aesar™ 4565906

 View more versions of this product

Catalog No. AA4565906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Tungsten (V) ethoxide 1,2-dimethoxyethane adduct
Yellow
5g
W(OC2H5)5·C4H10O2
UN1993
Liquid
Moisture Sensitive
<0°C
C10H25O5W·C4H10O2
MFCD16294494
499.26
99%

Chemical Identifiers

C10H25O5W·C4H10O2
MFCD16294494
Safety and Handling

Safety and Handling

GHS H Statement
H225-H360-H332-H315-H319
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P201-P309-P310
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents