Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$806.58 - $806.58

Chemical Identifiers

Chemical Name or Material Tungsten sputtering target
CAS 7440-33-7
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41641KS

 
alfa aesar™
41641KS
1 each Each for $806.58
Description & Specifications

Specifications

Chemical Name or Material Tungsten sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00011461
CAS 7440-33-7
Quantity 1 each