Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

$400.80 - $400.80

Chemical Identifiers

Chemical Name or Material Tungsten sputtering target
CAS 7440-33-7
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41640KS

  SDS
alfa aesar™
41640KS
1 each Each for $400.80
Description & Specifications

Specifications

Assay 99.95%
Assay Percent Notes (metals basis)
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011461
TSCA TSCA
CAS 7440-33-7
Quantity 1 each
Chemical Name or Material Tungsten sputtering target