Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$667.50 - $667.50

Chemical Identifiers

Chemical Name or Material Tungsten sputtering target
CAS 7440-33-7
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41639KS

  SDS
alfa aesar™
41639KS
1 each Each for $667.50
Description & Specifications

Specifications

MDL Number MFCD00011461
Assay Percent Notes (metals basis)
TSCA TSCA
Quantity 1 each
CAS 7440-33-7
Chemical Name or Material Tungsten sputtering target
Assay 99.95%
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick