Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

$808.12 - $808.12

Chemical Identifiers

Chemical Name or Material Tungsten sputtering target
CAS 7440-33-7
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41638KS

 
alfa aesar™
41638KS
1 each Each for $808.12
Description & Specifications

Specifications

Chemical Name or Material Tungsten sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00011461
CAS 7440-33-7
Quantity 1 each