missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.97% (metals basis)

Manufacturer:  Alfa Aesar™ 4425122

 View more versions of this product

Catalog No. AA4425122



Specifications

Specifications

Tungsten slug
100g
7440-33-7
W
4+ ion
[W]
183.84
CHEBI:27998
99.97%
≈3.9g/slug
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
(metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
≈3.9g/slug

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents