missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten rod, 6.3mm dia., annealed, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar™ 10406BS

 View more versions of this product

Catalog No. AA10406BS



Specifications

Specifications

Tungsten
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Rod, 6.3mm (0.25 in.) dia.
10cm
Silver Gray rod
W
4+ ion
[W]
183.84
CHEBI:27998

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

Documents