missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten rod, 5.0mm (0.20 in.) dia., annealed, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 44476BS

 View more versions of this product

Catalog No. AA44476BS



Specifications

Specifications

Tungsten rod
10cm
7440-33-7
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
99.95%
≈3.7g/cm
5.0mm (0.20 in.) dia., Annealed, Temper: soft
(metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Rod

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents