missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten rod, 5.0mm (0.20 in.) dia., annealed, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 44476BW

 View more versions of this product

Catalog No. AA44476BW



Specifications

Specifications

Tungsten rod
50cm
7440-33-7
W
4+ ion
[W]
183.84
CHEBI:27998
99.95%
≈3.7g/cm
5.0mm (0.20 in.) dia., Annealed, Temper: soft
(metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Rod, ≈3.7g/cm,Temper: soft

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents