Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten rod, 4.0mm (0.16 in.) dia., Puratronic™, 99.96% (metals basis)

Chemical Identifiers

Chemical Name or Material Tungsten rod
CAS 7440-33-7
Name Note 4.0mm (0.16 in.) dia.
Purity Grade Puratronic™
MDL Number MFCD00011461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11446DM

 
alfa aesar™
11446DM
100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11446HR

 
alfa aesar™
11446HR
5 x 100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tungsten rod
TSCA TSCA
Name Note 4.0mm (0.16 in.) dia.
Purity Grade Puratronic™
Physical Form Rod
Assay 99.96%
Assay Percent Notes (metals basis)
MDL Number MFCD00011461
CAS 7440-33-7
Weight ≈2.42g/cm
Quantity 100mm