missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten powder, APS <1 micron, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 44210A3

 View more versions of this product

Catalog No. AA44210A3



Specifications

Specifications

Tungsten powder
7440-33-7
W
UN3089
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Powder
99.95%
2kg
(metals basis)
MFCD00011461
4+ ion
[W]
183.84
CHEBI:27998
APS <1 Micron
Gray

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-143-9

RTECSNumber : YO7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents