missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten powder, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 10402A3

 View more versions of this product

Catalog No. AA10402A3



Specifications

Specifications

Tungsten
7440-33-7
99.9% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Powder
2kg
Light Gray powder
W
4+ ion
[W]
183.84
CHEBI:27998
Gray

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-143-9

Documents