missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten powder, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1040222

 View more versions of this product

Catalog No. AA1040222



Specifications

Specifications

Tungsten
7440-33-7
99.9% (Metals basis)
MFCD00011461
4+ ion, ion, powder, tungsten, elemental, tungsten, insoluble compounds, tungsten, soluble compounds, unii-v9306cxo6g, va, wolfram, wolframium
[W]
183.84
CHEBI:27998
Gray
100g
Light Gray powder
W
-100 Mesh
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Powder

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion, ion, powder, tungsten, elemental, tungsten, insoluble compounds, tungsten, soluble compounds, unii-v9306cxo6g, va, wolfram, wolframium
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228-H500

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-143-9

RTECSNumber : YO7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents