Please login to your online account to display your discounted pricing

Tungsten, plasma standard solution, Specpure™, W 10,000μg/ml, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tungsten, plasma standard solution, Specpure™
Molecular Formula W in 5% HNO3/tr. HF
MDL Number MFCD00011461
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA35772AD

  SDS
alfa aesar
35772AD
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35772AE

  SDS
alfa aesar
35772AE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35772AP

  SDS
alfa aesar
35772AP
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50mL
Odor Acidic
Health Hazard 2 GHS H Statement
H314-H302-H312-H332;
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Concentration W 10,000μg/ml
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
TSCA TSCA
Molecular Formula W in 5% HNO3/tr. HF
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
MDL Number MFCD00011461
Health Hazard 1 Danger
Chemical Name or Material Tungsten, plasma standard solution, Specpure™
UN Number UN3264
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H302-H312-H332;
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.