missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten(IV) Selenide, 99.8% (Metal Basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1308418

 View more versions of this product

Catalog No. AA1308418



Specifications

Specifications

Tungsten(IV) Selenide
99.8%, metal basis
MFCD00049703
Insoluble in water.
[Se]=[W]=[Se]
341.782
341.77
9.32
12067-46-8
Se2W
bis selanylidene tungsten
ROUIDRHELGULJS-UHFFFAOYSA-N
bis(selanylidene)tungsten
82910
10-20μm powder
50g

Chemical Identifiers

12067-46-8
341.782
ROUIDRHELGULJS-UHFFFAOYSA-N
82910
[Se]=[W]=[Se]
Se2W
MFCD00049703
bis selanylidene tungsten
bis(selanylidene)tungsten
Safety and Handling

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : DOT Class: 9; Packaging Group: III

EINECSNumber : 235-078-7

RTECSNumber : YO7714000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents