missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten(IV) Selenide, 99.8% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1308409

 View more versions of this product

Catalog No. AA1308409



Specifications

Specifications

Tungsten(IV) Selenide
99.8%, metal basis
MFCD00049703
ROUIDRHELGULJS-UHFFFAOYSA-N
bis(selanylidene)tungsten
82910
Powder, 10μm
10g
12067-46-8
Se2W
tungsten selenide, tungsten diselenide, tungsten selenide wse2, tungsten iv selenide, bis selanylidene tungsten, diselanylidenetungsten, tungsten diselenide, crystal
[Se]=[W]=[Se]
341.782
341.77
9.32

Chemical Identifiers

12067-46-8
341.782
ROUIDRHELGULJS-UHFFFAOYSA-N
82910
[Se]=[W]=[Se]
Se2W
MFCD00049703
tungsten selenide, tungsten diselenide, tungsten selenide wse2, tungsten iv selenide, bis selanylidene tungsten, diselanylidenetungsten, tungsten diselenide, crystal
bis(selanylidene)tungsten
Safety and Handling

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

Toxic by inhalation and if swallowed. Danger of cumulative effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

DOTInformation : DOT Class: 9

EINECSNumber : 235-078-7

Documents