Please login to your online account to display your discounted pricing

Tungsten ICP Standard, 1mL = 1mg W (1,000ppm W)W in 2% HNO3 / 1% HF, Ricca Chemical

$154.79 - $154.79

Chemical Identifiers

Chemical Name or Material Tungsten ICP Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 3.37
CAS Max % 3.5
CAS 7664-39-3
CAS Min % 1.21
CAS Max % 1.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PW1KN-100

 
ricca chemical company
RIPW1KN100
Poly natural 100mL Each for $154.79

PW1KN-500

 
ricca chemical company
RIPW1KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg W (1, 000ppm W)W in 2% HNO3 / 1% HF
Chemical Name or Material Tungsten ICP Standard
Shelf Life 18 months
CAS 7732-18-5
CAS 7440-33-7
CAS 7697-37-2
CAS Max % 1.26
CAS Max % 0.1
CAS Min % 93.0
CAS Min % 3.37
CAS Max % 97.0
CAS Min % 0.09
CAS Min % 1.21
CAS Max % 3.5
Packaging Poly natural
CAS 7664-39-3
Safety and Handling

18 months