missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten ICP Standard, 1mL = 10mg W (10,000ppm W), Ricca Chemical

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PW10KN-50
View Documents
Ricca Chemical Company
RIPW10KN50
50mL Poly natural N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PW10KN-100
View Documents
Ricca Chemical Company
RIPW10KN100
100mL Poly natural N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7440-33-7
183.84
23964
tungsten
W
WFKWXMTUELFFGS-UHFFFAOYSA-N
CHEBI:27998
[W]

Specifications

7440-33-7,7664-39-3,7664-39-3,7440-33-7
85.0,11.54,4.27,0.85
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Liquid
10,000ppm W
Colorless
Tungsten ICP Standard
82.0,11.09,4.1,0.82
W
[W]
183.84
CHEBI:27998
Poly natural
1 mL = 10 mg W
50mL