Learn More
Tungsten hexacarbonyl, 99%, Thermo Scientific Chemicals
Tungsten hexacarbonyl, 99%, C6O6W, CAS Number-14040-11-0, 250g, 175.0 deg.C, Yellow to Beige, 237-880-2, 98.5% min., 06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51, 351.91, GHS Signal Word: Danger, GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Supplier: Thermo Scientific Chemicals 221040500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
175.0°C | |
150.0°C | |
14040-11-0 | |
50 g | |
C6O6W | |
MFCD00011462 | |
Solubility in water: insoluble. Other solubilities: soluble in organic solvents | |
351.91 | |
Authentic |
Tungsten hexacarbonyl | |
Yellow to Beige | |
Fine Crystalline Powder | |
99% | |
W(CO6) | |
06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51 | |
351.91 | |
99% | |
Glass bottle |
Chemical Identifiers
14040-11-0 | |
C6O6W | |
351.91 | |
MFCD00011462 |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comforta
GHS Signal Word: Danger
EINECSNumber : 237-880-2
RUO – Research Use Only