Please login to your online account to display your discounted pricing

Tungsten hexacarbonyl, 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC221042500

 View more versions of this product

Catalog No. AC221042500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Formula Weight 351.91
Quantity 250g
Color Light yellow to beige
Fieser 06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 98.5% min.
Chemical Name or Material Tungsten hexacarbonyl
Assay Percent Range 99%
Molecular Formula C6O6W
Linear Formula W(CO6)
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00011462
CAS 14040-11-0
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.

Specifications

Chemical Name or Material Tungsten hexacarbonyl
Assay Percent Range 99%
Molecular Formula C6O6W
Linear Formula W(CO6)
Formula Weight 351.91
MDL Number MFCD00011462
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.

SDS & Safety Documents