missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten gauze, 150 mesh woven from 0.02mm (0.0008 in.) dia. wire

Manufacturer:  Alfa Aesar™ 47167FL

 View more versions of this product

Catalog No. AA47167FL



Specifications

Specifications

Tungsten gauze
150 mesh woven from 0.02mm (0.0008 in.) dia. wire
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
75 x 75mm
7440-33-7
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire Cloth

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

RTECSNumber : YO7175000

TSCA : TSCA

SDS Documents